Learn More
Research Products International Corp RPI Moxalactam, Sodium Salt, 500 Milligrams
SDP
Catalog No. 501978004
Encompass_Preferred
Moxalactam, Sodium Salt, also known as latamoxef sodium, is a broad-spectrum antibiotic belonging to the cephalosporin class. It is derived from 7-aminocephalosporanic acid and has potent activity against both gram-negative and gram-positive bacteria.
Moxalactam, Sodium Salt, works by inhibiting bacterial cell wall synthesis, leading to the disruption of the bacterial cell wall and ultimately causing cell death.
Its broad-spectrum activity makes it effective against various bacteria, including those resistant to other antibiotics.
Catalog No. 50-197-8004 Supplier Research Products International Corp Supplier No. M825000.5
Moxalactam, Sodium Salt, also known as latamoxef sodium, is a broad-spectrum antibiotic belonging to the cephalosporin class. It is derived from 7-aminocephalosporanic acid and has potent activity against both gram-negative and gram-positive bacteria.
Moxalactam, Sodium Salt, works by inhibiting bacterial cell wall synthesis, leading to the disruption of the bacterial cell wall and ultimately causing cell death.
Its broad-spectrum activity makes it effective against various bacteria, including those resistant to other antibiotics.