Learn More
Research Products International Corp Moxalactam, Sodium Salt, 1 Gram
SDP
Catalog No. 50997929
Encompass_Preferred
Moxalactam, Sodium Salt, also known as latamoxef sodium, is a broad-spectrum antibiotic belonging to the cephalosporin class. It is derived from 7-aminocephalosporanic acid and has potent activity against both gram-negative and gram-positive bacteria.
Moxalactam, Sodium Salt, works by inhibiting bacterial cell wall synthesis, leading to the disruption of the bacterial cell wall and ultimately causing cell death.
Its broad-spectrum activity makes it effective against various bacteria, including those resistant to other antibiotics.
Catalog No. 50-997-929 Supplier Research Products International Corp Supplier No. M825001.0
Moxalactam, Sodium Salt, also known as latamoxef sodium, is a broad-spectrum antibiotic belonging to the cephalosporin class. It is derived from 7-aminocephalosporanic acid and has potent activity against both gram-negative and gram-positive bacteria.
Moxalactam, Sodium Salt, works by inhibiting bacterial cell wall synthesis, leading to the disruption of the bacterial cell wall and ultimately causing cell death.
Its broad-spectrum activity makes it effective against various bacteria, including those resistant to other antibiotics.