Learn More
Aviva Systems Biology Corp MOXD1 Antibody - C-terminal region (ARP62346_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62346_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MOXD1. It was validated on Western Blot. . Alternate names: DKFZp564G202, MOX, PRO5780, dJ248E1.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-287-655