Learn More
TARGETMOL CHEMICALS INC MOXIDECTIN 50MG
SDP
Catalog No. 502577452
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Moxidectin (CL301423) is an anthelmintic drug which kills parasitic worms and is used for the prevention and control of heartworm and intestinal worms. It can be found in treatments prescribed for animals such as dogs cats horses cattle and sheep. Application methods for moxidectin vary by treatment and include oral topical and injectable solutions. Cydectin Pour On is a trade name for a formulation for use on cattle and red deer. purity: 99%
Catalog No. 50-257-7452 Supplier TARGETMOL CHEMICALS INC Supplier No. T115750MG
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Moxidectin (CL301423) is an anthelmintic drug which kills parasitic worms and is used for the prevention and control of heartworm and intestinal worms. It can be found in treatments prescribed for animals such as dogs cats horses cattle and sheep. Application methods for moxidectin vary by treatment and include oral topical and injectable solutions. Cydectin Pour On is a trade name for a formulation for use on cattle and red deer. purity: 99%