Learn More
Apexbio Technology LLC MOXIFLOXACIN-D4-1MG
SDP
Catalog No. 5000570257
Encompass_Preferred
Moxifloxacin-d4 (CAS No 2596386-23-9) is a deuterated derivative used in biomedical research primarily focused on bacteriology and pharmacokinetics Originating from synthetic modifications of the quinolone antibiotic moxifloxacin it functions by inhibiting bacterial DNA gyrase and topoisomerase IV essential enzymes for DNA replication This inhibition disrupts DNA processes leading to bacterial cell death Moxifloxacin-d4 exhibits potent antibacterial activity in vitro typically in the nanomolar to low micromolar range Its applications often include various cell models and animal studies to understand pharmacokinetics bioavailability and drug interaction Additionally it serves in drug screening processes to evaluate efficacy and metabolic pathways Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70257 Supplier Apexbio Technology LLC Supplier No. SL10511
Moxifloxacin-d4 (CAS No 2596386-23-9) is a deuterated derivative used in biomedical research primarily focused on bacteriology and pharmacokinetics Originating from synthetic modifications of the quinolone antibiotic moxifloxacin it functions by inhibiting bacterial DNA gyrase and topoisomerase IV essential enzymes for DNA replication This inhibition disrupts DNA processes leading to bacterial cell death Moxifloxacin-d4 exhibits potent antibacterial activity in vitro typically in the nanomolar to low micromolar range Its applications often include various cell models and animal studies to understand pharmacokinetics bioavailability and drug interaction Additionally it serves in drug screening processes to evaluate efficacy and metabolic pathways Experimental concentrations typically depend on the specific research design