Learn More
Medchemexpress LLC Moxifloxacin-d4 hydrochloride | C21H21D4ClFN3O4 | 1 MG
SDP
Catalog No. 5000434949
Encompass_Preferred
Moxifloxacin-d4 hydrochloride is the hydrochloride salt form of deuterium labeled Moxifloxacin. It is an orally active antimicrobial that can be used in the research of acute bacterial sinusitis, acute bacterial exacerbations of chronic bronchitis, and community-acquired pneumonia.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-34949 Supplier Medchemexpress LLC Supplier No. HY66011AS61MG
Add to Cart
Add to Cart
Moxifloxacin-d4 hydrochloride is the hydrochloride salt form of deuterium labeled Moxifloxacin. It is an orally active antimicrobial that can be used in the research of acute bacterial sinusitis, acute bacterial exacerbations of chronic bronchitis, and community-acquired pneumonia.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS