Learn More
Apexbio Technology LLC Moxifloxacin HCl 186826-86-8 200mg
SDP
Catalog No. 5000093531
Encompass_Preferred
Moxifloxacin HCl (186826-86-8) is a small-molecule inhibitor targeting bacterial DNA gyrase (topoisomerase II) and topoisomerase IV It is designed to inhibit these enzymes thereby disrupting essential bacterial DNA replication transcription and repair processes Moxifloxacin HCl exerts its biological activity primarily through inhibition of DNA gyrase and topoisomerase IV It demonstrates antibacterial activity against both Gram-positive and Gram-negative bacteria including ciprofloxacin-resistant strains with MIC90 values for ciprofloxacin-susceptible strains up to 0 6 g/mL Additionally moxifloxacin inhibits human serum paraoxonase-1 (hPON1) with a Ki value of 2 641 0 004 mM Based on these pharmacological properties Moxifloxacin HCl holds research potential in studying antibacterial mechanisms and enzyme-inhibitor interactions
Catalog No. 50-000-93531 Supplier Apexbio Technology LLC Supplier No. A5323200
Moxifloxacin HCl (186826-86-8) is a small-molecule inhibitor targeting bacterial DNA gyrase (topoisomerase II) and topoisomerase IV It is designed to inhibit these enzymes thereby disrupting essential bacterial DNA replication transcription and repair processes Moxifloxacin HCl exerts its biological activity primarily through inhibition of DNA gyrase and topoisomerase IV It demonstrates antibacterial activity against both Gram-positive and Gram-negative bacteria including ciprofloxacin-resistant strains with MIC90 values for ciprofloxacin-susceptible strains up to 0 6 g/mL Additionally moxifloxacin inhibits human serum paraoxonase-1 (hPON1) with a Ki value of 2 641 0 004 mM Based on these pharmacological properties Moxifloxacin HCl holds research potential in studying antibacterial mechanisms and enzyme-inhibitor interactions