Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moxifloxacin Hydrochloride Monohydrate 98.0+%, TCI America™
SDP

Catalog No. M2479100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2479100MG 100 mg
M24791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2479100MG Supplier TCI America Supplier No. M2479100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 192927-63-2
Molecular Formula C21H27ClFN3O5
Molecular Weight (g/mol) 455.91
MDL Number MFCD09836237
InChI Key SKZIMSDWAIZNDD-UHFFFAOYNA-N
Synonym 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aS,7aS)-octahydro-1H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic Acid Hydrochloride
PubChem CID 9890250
IUPAC Name 1-cyclopropyl-6-fluoro-8-methoxy-7-{octahydro-1H-pyrrolo[3,4-b]pyridin-6-yl}-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrate hydrochloride
SMILES O.Cl.COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CC2CCCNC2C1)C1CC1

Specifications

Melting Point 240°C
Quantity 100 mg
Formula Weight 437.90
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Moxifloxacin Hydrochloride Monohydrate
RTECSNumber VB1983750
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.