Learn More
Cayman Chemical Moxifloxacin (hydrochloride), Purity: ≥95%, CAS: 186826-86-8, 50 mg
SDP
Catalog No. 502971444
Encompass_Preferred
A fluoroquinolone antibiotic; active against 390 clinical isolates of various bacteria (MIC90s = ≤0.25 µg/ml), as well as clinical isolates of methicillin-susceptible and -resistant S. aureus (MIC90s = 0.12 and 2 µg/ml, respectively); an inhibitor of E. coli DNA gyrase that is selective for DNA gyrase over E. coli topoisomerase IV (IC50s = 0.51 and 38.8 mg/L, respectively, in cell-free assays); prevents S. aureus- and P. aeruginosa-induced increases in BALF neutrophil infiltration and reduces S. aureus- and P. aeruginosa-induced increases in lung CXCL1 and IL-1β levels in mouse models of bacterial pneumonia at 100 mg/kg twice per day for two days; decreases the number of lung and spleen CFUs in a mouse model of systemic M. tuberculosis infection at 100 mg/kg
Catalog No. 50-297-1444 Supplier Cayman Chemical Supplier No. 1483050 MG
A fluoroquinolone antibiotic; active against 390 clinical isolates of various bacteria (MIC90s = ≤0.25 µg/ml), as well as clinical isolates of methicillin-susceptible and -resistant S. aureus (MIC90s = 0.12 and 2 µg/ml, respectively); an inhibitor of E. coli DNA gyrase that is selective for DNA gyrase over E. coli topoisomerase IV (IC50s = 0.51 and 38.8 mg/L, respectively, in cell-free assays); prevents S. aureus- and P. aeruginosa-induced increases in BALF neutrophil infiltration and reduces S. aureus- and P. aeruginosa-induced increases in lung CXCL1 and IL-1β levels in mouse models of bacterial pneumonia at 100 mg/kg twice per day for two days; decreases the number of lung and spleen CFUs in a mouse model of systemic M. tuberculosis infection at 100 mg/kg