Learn More
Medchemexpress LLC Moxisylyte (hydrochloride) | 964-52-3 | 99.7% | 1 ML
SDP
Catalog No. 5000412670
Encompass_Preferred
Moxisylyte (hydrochloride) is a selective α1A-adrenergic receptor antagonist that competitively antagonizes norepinephrine activation. It relaxes penile cavernous smooth muscle and aids erectile function, also improving dysuria and reducing residual urine volume in patients with multiple system atrophy. It is intended for research related to erectile dysfunction and multiple system atrophy.
  • Acts as a competitive α-adrenergic receptor antagonist
  • Exerts concentration-dependent relaxant effect on isolated human corpus cavernosum tissues
  • Applicable to research related to erectile dysfunction
  • Applicable to research related to multiple system atrophy
Catalog No. 50-004-12670 Supplier Medchemexpress LLC Supplier No. HYB143510MM/1ML
Moxisylyte (hydrochloride) is a selective α1A-adrenergic receptor antagonist that competitively antagonizes norepinephrine activation. It relaxes penile cavernous smooth muscle and aids erectile function, also improving dysuria and reducing residual urine volume in patients with multiple system atrophy. It is intended for research related to erectile dysfunction and multiple system atrophy.
  • Acts as a competitive α-adrenergic receptor antagonist
  • Exerts concentration-dependent relaxant effect on isolated human corpus cavernosum tissues
  • Applicable to research related to erectile dysfunction
  • Applicable to research related to multiple system atrophy