Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Moxonidine hydrochloride | 75536-04-8 | 99.8% | 1 ML
SDP

Catalog No. 5000406487
Encompass_Preferred

Moxonidine hydrochloride | 75536-04-8 | 99.8% | 1 ML

Catalog No. 50-004-06487 Supplier Medchemexpress LLC Supplier No. HYB0374A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Moxonidine hydrochloride | 75536-04-8 | 99.8% | 1 ML

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.