Learn More
Abcam Mo x onidine hydrochloride, I1 Imidazoline receptor and alpha2 adrenergic agonist, 10MG
Catalog No. 502707267
Encompass_Preferred
MW 278.14 g/mol, Purity >99%. I₁ Imidazoline receptor and α₂ adrenergic agonist. Displays 4-fold greater affinity at I₁ receptors than α₂ receptors. Centrally active. Displays antihypertensive action.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7267 Supplier Abcam Supplier No. AB12078910MG
MW 278.14 g/mol, Purity >99%. I₁ Imidazoline receptor and α₂ adrenergic agonist. Displays 4-fold greater affinity at I₁ receptors than α₂ receptors. Centrally active. Displays antihypertensive action.
The product is subject to the following: Abcam Restricted Use Statement