Learn More
Medchemexpress LLC Moz-in-2 | 2055397-88-9 | MFCD31617713 | 99.6% | 372.37 g/mol | C17H13FN4O3S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10205925MG
Encompass_Preferred
Moz-in-2 is a small-molecule inhibitor of the histone acetyltransferase MOZ (KAT6A), reported with an IC50 of 125 μM. It is supplied as a high-purity solid for research use, with molecular formula C17H13FN4O3S and molecular weight 372.37 g/mol. Handle and store per supplier guidance for long-term stability.
  • Inhibits MOZ (KAT6A) with IC50 = 125 μM.
  • High purity (≈99.6%).
  • Solid form suitable for biochemical and cellular assays.
  • Provided in small research quantities (25 mg).
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
Catalog No. 50-002-47751