Learn More
TARGETMOL CHEMICALS INC MOZAVAPTAN 50MG
SDP
Catalog No. 502579453
Encompass_Preferred
Also available in 10 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mozavaptan (OPC-31260) is a competitive vasopressin receptor antagonist for both V1 and V2 receptors with IC50 of 1.2 uM and 14 nM respectively. purity: 99%
Catalog No. 50-257-9453 Supplier TARGETMOL CHEMICALS INC Supplier No. T628850MG
Also available in 10 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mozavaptan (OPC-31260) is a competitive vasopressin receptor antagonist for both V1 and V2 receptors with IC50 of 1.2 uM and 14 nM respectively. purity: 99%