Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mozavaptan (hydrochloride) | 138470-70-9 | MFCD11111965 | 99.9% | 464.0 g/mol | C27H30ClN3O2 | 10MM 1ML
SDP

Catalog No. 5000238880
Encompass_Preferred

Mozavaptan hydrochloride (OPC-31260) is the hydrochloride salt of a selective vasopressin V2 receptor antagonist. This product is supplied as a 10 mM solution in DMSO in a 1 mL vial for laboratory research use only.

  • Ready-to-use 10 mM solution in DMSO.
  • Supplied in a 1 mL sealed vial.
  • High purity (99.9%).
  • Molecular formula C27H30ClN3O2; molecular weight 464.0 g/mol.
  • Recommended storage: 4°C sealed; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • For research use only; not for human or diagnostic applications.

Catalog No. 50-002-38880 Supplier Medchemexpress LLC Supplier No. HY12359310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mozavaptan hydrochloride (OPC-31260) is the hydrochloride salt of a selective vasopressin V2 receptor antagonist. This product is supplied as a 10 mM solution in DMSO in a 1 mL vial for laboratory research use only.

  • Ready-to-use 10 mM solution in DMSO.
  • Supplied in a 1 mL sealed vial.
  • High purity (99.9%).
  • Molecular formula C27H30ClN3O2; molecular weight 464.0 g/mol.
  • Recommended storage: 4°C sealed; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • For research use only; not for human or diagnostic applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.