Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mozavaptan hydrochloride | 138470-70-9 | MFCD11111965 | 99.9% | 464.0 g/mol | C27H30ClN3O2 | 50MG
SDP

Supplier:  Medchemexpress LLC HY12359350MG

Encompass_Preferred

Mozavaptan hydrochloride (CAS 138470-70-9) is a benzazepine-derived, selective vasopressin V2 receptor antagonist supplied as a high-purity research chemical for biochemical and pharmacological studies. It is provided as a solid with defined storage conditions and is suitable as a reference standard or for in vitro pharmacology.

  • Selective vasopressin V2 receptor antagonist
  • High purity (99.9%) solid suitable for research
  • Molecular formula C27H30ClN3O2; molecular weight 464.0 g/mol
  • Available in small pack sizes for experimental use
  • Store sealed, away from moisture at 4°C; in solvent: -80°C (6 months) or -20°C (1 month)
  • CAS number 138470-70-9 for unambiguous identification

Catalog No. 50-002-37692


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.