Learn More
Medchemexpress LLC Mozavaptan hydrochloride | 138470-70-9 | MFCD11111965 | 99.9% | 464.0 g/mol | C27H30ClN3O2 | 5MG
SDP
Catalog No. 5000237535
Encompass_Preferred
Mozavaptan hydrochloride (OPC-31260 hydrochloride) is a benzazepine-derived, potent and selective vasopressin V2 receptor antagonist (reported IC50 ≈14 nM) used as a research chemical for studies of vasopressin signaling and conditions such as hyponatremia and SIADH. It is supplied as a white to off-white solid and is commonly available in small milligram quantities or as a DMSO solution for in vitro and pharmacology experiments. Molecular formula C27H30ClN3O2; molecular weight 464.0 g/mol; CAS 138470-70-9.
  • High purity suitable for research (≈99.9%)
  • Potent V2 receptor antagonist activity for pharmacology studies
  • Available in solid and DMSO solution formats
  • Compact packaging for small-scale experiments
  • Standard identifiers provided for traceability
Catalog No. 50-002-37535 Supplier Medchemexpress LLC Supplier No. HY1235935MG
Mozavaptan hydrochloride (OPC-31260 hydrochloride) is a benzazepine-derived, potent and selective vasopressin V2 receptor antagonist (reported IC50 ≈14 nM) used as a research chemical for studies of vasopressin signaling and conditions such as hyponatremia and SIADH. It is supplied as a white to off-white solid and is commonly available in small milligram quantities or as a DMSO solution for in vitro and pharmacology experiments. Molecular formula C27H30ClN3O2; molecular weight 464.0 g/mol; CAS 138470-70-9.
  • High purity suitable for research (≈99.9%)
  • Potent V2 receptor antagonist activity for pharmacology studies
  • Available in solid and DMSO solution formats
  • Compact packaging for small-scale experiments
  • Standard identifiers provided for traceability