Learn More
Medchemexpress LLC Mozavaptan hydrochloride | 138470-70-9 | MFCD11111965 | 99.9% | 464.00 | C27H30ClN3O2 | 100MG
SDP
Catalog No. 5000237662
Encompass_Preferred
Mozavaptan hydrochloride is a benzazepine-derived, selective vasopressin V2 receptor antagonist supplied as the hydrochloride salt for research applications. It antagonizes arginine vasopressin's antidiuretic action and is commonly used to model and study disorders of water balance such as hyponatremia and SIADH. The material is provided with specified purity and recommended storage conditions for laboratory use.
  • Potent V2 receptor antagonist (IC50 ≈ 14 nM).
  • Approximately 85-fold selectivity for V2 over V1 receptor.
  • High purity for research use (99.89%).
  • Available in multiple pack sizes, including 100 mg.
  • Stable when stored sealed at 4°C; in solvent: -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-37662 Supplier Medchemexpress LLC Supplier No. HY123593100MG
Mozavaptan hydrochloride is a benzazepine-derived, selective vasopressin V2 receptor antagonist supplied as the hydrochloride salt for research applications. It antagonizes arginine vasopressin's antidiuretic action and is commonly used to model and study disorders of water balance such as hyponatremia and SIADH. The material is provided with specified purity and recommended storage conditions for laboratory use.
  • Potent V2 receptor antagonist (IC50 ≈ 14 nM).
  • Approximately 85-fold selectivity for V2 over V1 receptor.
  • High purity for research use (99.89%).
  • Available in multiple pack sizes, including 100 mg.
  • Stable when stored sealed at 4°C; in solvent: -80°C (6 months) or -20°C (1 month).