Learn More
Medchemexpress LLC Mozavaptan hydrochloride | 138470-70-9 | MFCD11111965 | >=98.0% | 464.00 g/mol | C27H30ClN3O2 | 10MG
SDP
Catalog No. 5000235069
Encompass_Preferred
Mozavaptan hydrochloride is a benzazepine-derived vasopressin V2 receptor antagonist used as a research reagent to study antidiuretic mechanisms and disorders such as hyponatremia and SIADH. It is provided as a high-purity hydrochloride salt for in vitro and in vivo pharmacology studies and analytical reference standards.
  • Potent, selective antagonist of vasopressin V2 receptor for pharmacological research.
  • Suitable for in vitro and in vivo studies of antidiuretic pathways.
  • Provided as a hydrochloride salt with defined molecular formula and weight.
  • Available in small pack sizes appropriate for screening and preclinical work.
  • Typically supplied at high purity suitable for analytical and biological assays.
Catalog No. 50-002-35069 Supplier Medchemexpress LLC Supplier No. HY12359310MG
Mozavaptan hydrochloride is a benzazepine-derived vasopressin V2 receptor antagonist used as a research reagent to study antidiuretic mechanisms and disorders such as hyponatremia and SIADH. It is provided as a high-purity hydrochloride salt for in vitro and in vivo pharmacology studies and analytical reference standards.
  • Potent, selective antagonist of vasopressin V2 receptor for pharmacological research.
  • Suitable for in vitro and in vivo studies of antidiuretic pathways.
  • Provided as a hydrochloride salt with defined molecular formula and weight.
  • Available in small pack sizes appropriate for screening and preclinical work.
  • Typically supplied at high purity suitable for analytical and biological assays.