Learn More
Cayman Chemical Mozavaptan (hydrochloride), Purity: ≥98%, CAS: 138470-70-9, 50 mg
SDP
Catalog No. 502965617
Encompass_Preferred
A vasopressin V2 receptor antagonist (Ki = 9.42 nM in HeLa cells expressing the human receptor); selective for vasopressin V2 over V1 receptors (IC50s = 14 and 1,200 nM, respectively) in radioligand binding assays using rat kidney and rat liver membranes, respectively; increases urine volume and decreases urine osmolality in rats from 10-30 mg/kg
Catalog No. 50-296-5617 Supplier Cayman Chemical Supplier No. 1666450 MG
A vasopressin V2 receptor antagonist (Ki = 9.42 nM in HeLa cells expressing the human receptor); selective for vasopressin V2 over V1 receptors (IC50s = 14 and 1,200 nM, respectively) in radioligand binding assays using rat kidney and rat liver membranes, respectively; increases urine volume and decreases urine osmolality in rats from 10-30 mg/kg