Learn More
Cayman Chemical MMP-13 Inhibitor, Purity: ≥98%, CAS: 544678-85-5, 10 mg
SDP
Catalog No. 502982525
Encompass_Preferred
A pyrimidine dicarboxamide that selectively inhibits MMP-13 (IC50 = 8 nM); blocks osterix-dependent calcification of matrices in limb bud cells during endochondral ossification
Catalog No. 50-298-2525 Supplier Cayman Chemical Supplier No. 1954010 MG
A pyrimidine dicarboxamide that selectively inhibits MMP-13 (IC50 = 8 nM); blocks osterix-dependent calcification of matrices in limb bud cells during endochondral ossification