Learn More
Cayman Chemical MMP-13 Inhibitor, Purity: ≥98%, CAS: 544678-85-5, 1 mg
SDP
Catalog No. 502983555
Encompass_Preferred
A pyrimidine dicarboxamide that selectively inhibits MMP-13 (IC50 = 8 nM); blocks osterix-dependent calcification of matrices in limb bud cells during endochondral ossification
Catalog No. 50-298-3555 Supplier Cayman Chemical Supplier No. 195401 MG
A pyrimidine dicarboxamide that selectively inhibits MMP-13 (IC50 = 8 nM); blocks osterix-dependent calcification of matrices in limb bud cells during endochondral ossification