Learn More
Cayman Chemical mP-13 InhIbItr 5mg
SDP
Catalog No. 502983418
Encompass_Preferred
A pyrimidine dicarboxamide that selectively inhibits MMP-13 (IC50 = 8 nM); blocks osterix-dependent calcification of matrices in limb bud cells during endochondral ossification
Catalog No. 50-298-3418 Supplier Cayman Chemical Supplier No. 195405 MG
Add to Cart
Add to Cart
A pyrimidine dicarboxamide that selectively inhibits MMP-13 (IC50 = 8 nM); blocks osterix-dependent calcification of matrices in limb bud cells during endochondral ossification