Learn More
Cayman Chemical mP-2 InhIbItr I 10mg
SDP
Catalog No. 502981930
Encompass_Preferred
A hydroxamate-based, long-chain fatty acid that reversibly inhibits MMP-2 (Ki = 1.6 µM); attenuates cancer cell migration and has been shown to preserve blood-brain barrier function in a rat model of pneumococcal meningitis
Catalog No. 50-298-1930 Supplier Cayman Chemical Supplier No. 1964410 MG
Add to Cart
Add to Cart
A hydroxamate-based, long-chain fatty acid that reversibly inhibits MMP-2 (Ki = 1.6 µM); attenuates cancer cell migration and has been shown to preserve blood-brain barrier function in a rat model of pneumococcal meningitis