Learn More
Cayman Chemical MMP-2 Inhibitor I, Purity: ≥98%, CAS: 10335-69-0, 5 mg
SDP
Catalog No. 502985563
Encompass_Preferred
A hydroxamate-based, long-chain fatty acid that reversibly inhibits MMP-2 (Ki = 1.6 µM); attenuates cancer cell migration and has been shown to preserve blood-brain barrier function in a rat model of pneumococcal meningitis
Catalog No. 50-298-5563 Supplier Cayman Chemical Supplier No. 196445 MG
A hydroxamate-based, long-chain fatty acid that reversibly inhibits MMP-2 (Ki = 1.6 µM); attenuates cancer cell migration and has been shown to preserve blood-brain barrier function in a rat model of pneumococcal meningitis