Learn More
Cayman Chemical mP-2 InhIbItr II 10mg
SDP
Catalog No. 502980304
Encompass_Preferred
An analog of SB-3CT that irreversibly inhibits MMP-2 (Ki = 2.4 µM); less potently inhibits MMP-1 and -7 (Kis = 45 and 379 µM, respectively); attenuates glucose-induced MMP-2 activity and expression, as well as subsequent apoptosis, in retinal endothelial cells
Catalog No. 50-298-0304 Supplier Cayman Chemical Supplier No. 1726710 MG
Add to Cart
Add to Cart
An analog of SB-3CT that irreversibly inhibits MMP-2 (Ki = 2.4 µM); less potently inhibits MMP-1 and -7 (Kis = 45 and 379 µM, respectively); attenuates glucose-induced MMP-2 activity and expression, as well as subsequent apoptosis, in retinal endothelial cells