Learn More
Cayman Chemical MMP-2 Inhibitor II, Purity: ≥98%, CAS: 869577-51-5, 5 mg
SDP
Catalog No. 502979641
Encompass_Preferred
An analog of SB-3CT that irreversibly inhibits MMP-2 (Ki = 2.4 µM); less potently inhibits MMP-1 and -7 (Kis = 45 and 379 µM, respectively); attenuates glucose-induced MMP-2 activity and expression, as well as subsequent apoptosis, in retinal endothelial cells
Catalog No. 50-297-9641 Supplier Cayman Chemical Supplier No. 172675 MG
An analog of SB-3CT that irreversibly inhibits MMP-2 (Ki = 2.4 µM); less potently inhibits MMP-1 and -7 (Kis = 45 and 379 µM, respectively); attenuates glucose-induced MMP-2 activity and expression, as well as subsequent apoptosis, in retinal endothelial cells