Learn More
Cayman Chemical mP-2mP-9 InhIbItr I 10mg
SDP
Catalog No. 502980820
Encompass_Preferred
A potent inhibitor of MMP-2 and MMP-9 (IC50s = 310 and 240 nM, respectively) that is active in vivo as well as in vitro
Catalog No. 50-298-0820 Supplier Cayman Chemical Supplier No. 2031510 MG
Add to Cart
Add to Cart
A potent inhibitor of MMP-2 and MMP-9 (IC50s = 310 and 240 nM, respectively) that is active in vivo as well as in vitro