Learn More
Cayman Chemical MMP-2/MMP-9 Inhibitor I, Purity: ≥99%, CAS: 193807-58-8, 25 mg
SDP
Catalog No. 502980823
Encompass_Preferred
A potent inhibitor of MMP-2 and MMP-9 (IC50s = 310 and 240 nM, respectively) that is active in vivo as well as in vitro
Catalog No. 50-298-0823 Supplier Cayman Chemical Supplier No. 2031525 MG
A potent inhibitor of MMP-2 and MMP-9 (IC50s = 310 and 240 nM, respectively) that is active in vivo as well as in vitro