Learn More
Cayman Chemical mP-2mP-9 InhIbItr II 1mg
SDP
Catalog No. 502936563
Encompass_Preferred
A dual inhibitor of MMP-2 and MMP-9 (IC50s = 17 and 30 nM, respectively); suppresses lung colonization of Lewis lung carcinoma cells and inhibits tumor-induced angiogenesis, tumor growth, and liver metastasis in mice
Catalog No. 50-293-6563 Supplier Cayman Chemical Supplier No. 243141 MG
Add to Cart
Add to Cart
A dual inhibitor of MMP-2 and MMP-9 (IC50s = 17 and 30 nM, respectively); suppresses lung colonization of Lewis lung carcinoma cells and inhibits tumor-induced angiogenesis, tumor growth, and liver metastasis in mice