Learn More
Cayman Chemical MMP-3 Fluorogenic Substrate (trifluoroacetate salt), Purity: ≥95%, 5 mg
SDP
Catalog No. 502917388
Encompass_Preferred
A fluorogenic substrate for MMP-3; Mca is released upon cleavage by MMP-3 and its fluorescence can be used to quantify MMP-3 activity
Catalog No. 50-291-7388 Supplier Cayman Chemical Supplier No. 375235 MG
A fluorogenic substrate for MMP-3; Mca is released upon cleavage by MMP-3 and its fluorescence can be used to quantify MMP-3 activity