Learn More
Cayman Chemical MMP-3 Inhibitor VIII, Purity: ≥95%, CAS: 208663-26-7, 1 mg
SDP
Catalog No. 502968129
Encompass_Preferred
A cell permeable sulfonamide-based hydroxamic acid that binds to the active site of MMP-3 (Ki = 23 nM) and prevents its activity; inhibits mouse macrophage metalloelastase MME/MMP-12 (IC50 = 13 nM)
Catalog No. 50-296-8129 Supplier Cayman Chemical Supplier No. 172461 MG
A cell permeable sulfonamide-based hydroxamic acid that binds to the active site of MMP-3 (Ki = 23 nM) and prevents its activity; inhibits mouse macrophage metalloelastase MME/MMP-12 (IC50 = 13 nM)