Learn More
Cayman Chemical mP-3 InhIbItr VIII 500ug
SDP
Catalog No. 502966634
Encompass_Preferred
A cell permeable sulfonamide-based hydroxamic acid that binds to the active site of MMP-3 (Ki = 23 nM) and prevents its activity; inhibits mouse macrophage metalloelastase MME/MMP-12 (IC50 = 13 nM)
Catalog No. 50-296-6634 Supplier Cayman Chemical Supplier No. 17246500 UG
A cell permeable sulfonamide-based hydroxamic acid that binds to the active site of MMP-3 (Ki = 23 nM) and prevents its activity; inhibits mouse macrophage metalloelastase MME/MMP-12 (IC50 = 13 nM)