Learn More
Selleck Chemical LLC MP-513 hydrobromide hydrate E4814-25MG
SDP
Catalog No. 5000042246
Encompass_Preferred
Teneligliptin Hydrobromide Hydrate(MP-513 hydrobromide hydrate) is a potent competitive and long-lasting inhibitor of DPP-4 It competitively inhibited rhDPP-4 human plasma and rat plasma with IC50 values of 0 889 1 75 and 1 35 nM respectively It suppresses high glucose-mediated mitochondrial dysfunction and can be used for treatment of type 2 diabetes
Catalog No. 50-000-42246 Supplier Selleck Chemical LLC Supplier No. E481425MG
Teneligliptin Hydrobromide Hydrate(MP-513 hydrobromide hydrate) is a potent competitive and long-lasting inhibitor of DPP-4 It competitively inhibited rhDPP-4 human plasma and rat plasma with IC50 values of 0 889 1 75 and 1 35 nM respectively It suppresses high glucose-mediated mitochondrial dysfunction and can be used for treatment of type 2 diabetes