Learn More
Cayman Chemical mP-9 InhIbItr I 500ug
SDP
Catalog No. 502972070
Encompass_Preferred
An inhibitor of MMP-9; decreases TACE activity (IC50 = 0.54 µM)
Catalog No. 50-297-2070 Supplier Cayman Chemical Supplier No. 15942500 UG
An inhibitor of MMP-9; decreases TACE activity (IC50 = 0.54 µM)