Learn More
Cayman Chemical mP-9 Q279R VarInthumnr 20ug
SDP
Catalog No. 502950412
Encompass_Preferred
Active recombinant C-terminal His-tagged protein expressed in HEK293 cells • Amino acids: 20-707 • MW: 77.8 kDa
Catalog No. 50-295-0412 Supplier Cayman Chemical Supplier No. 3256020 UG
Active recombinant C-terminal His-tagged protein expressed in HEK293 cells • Amino acids: 20-707 • MW: 77.8 kDa