Learn More
Roche Diagnostics MagNA Pure 96 Processing Cartridge, 36 cartridges
Catalog No. 50588366 Shop All Roche Diagnostics Products
Encompass
MagNA Pure 96 disposables are nuclease free according to current Quality Control procedures. They are manufactured from material that is inert to chemical leaching at low and elevated temperatures, that could interfere with downstream applications.
Catalog No. 50-588-366 Supplier Roche Diagnostics Supplier No. 06241603001
MagNA Pure 96 disposables are nuclease free according to current Quality Control procedures. They are manufactured from material that is inert to chemical leaching at low and elevated temperatures, that could interfere with downstream applications.