Learn More
Roche Diagnostics MagNA Pure 96 System Fluid (External)
Catalog No. NC1848246 Shop All Roche Diagnostics Products
Encompass
For rinsing the reagent head of the MagNA Pure 96 Instrument during a purification run. When the external configuration of system fluid and liquid waste are chosen, it is recommended to place it on a table that can also hold the external containers.
Catalog No. NC1848246 Supplier Roche Diagnostics Supplier No. 06640729001
For rinsing the reagent head of the MagNA Pure 96 Instrument during a purification run. When the external configuration of system fluid and liquid waste are chosen, it is recommended to place it on a table that can also hold the external containers.