Learn More
Cayman Chemical mP-9mP-13 InhIbItr I 500ug
SDP
Catalog No. 502985456
Encompass_Preferred
A cell-permeable inhibitor of MMPs that most potently inhibits MMP-9 and MMP-13 (IC50s = 0.9 nM for both); less effectively inhibits MMP-1, MMP-3, and MMP-7 (IC50s = 43, 23, and 920 nM, respectively)
Catalog No. 50-298-5456 Supplier Cayman Chemical Supplier No. 21265500 UG
Add to Cart
Add to Cart
A cell-permeable inhibitor of MMPs that most potently inhibits MMP-9 and MMP-13 (IC50s = 0.9 nM for both); less effectively inhibits MMP-1, MMP-3, and MMP-7 (IC50s = 43, 23, and 920 nM, respectively)