Learn More
Medchemexpress LLC HY-19794 5mg Medchemexpress, MP-A08 CAS:219832-49-2 Purity:>98%
SDP
Catalog No. 502082186
Encompass_Preferred
Medchemexpress, HY-19794 5mg MP-A08 CAS:219832-49-2 MP-A08 is a highly selective ATP competitive sphingosine kinase (SPHK1) inhibitor that targets both SphK1 and SphK2 with Ki values of 6.9 ± 0.8 μM and 27 ± 3 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2186 Supplier Medchemexpress LLC Supplier No. HY197945MG
Medchemexpress, HY-19794 5mg MP-A08 CAS:219832-49-2 MP-A08 is a highly selective ATP competitive sphingosine kinase (SPHK1) inhibitor that targets both SphK1 and SphK2 with Ki values of 6.9 ± 0.8 μM and 27 ± 3 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.