Learn More
Medchemexpress LLC MP-Ala-Ala-PAB | 1949793-47-8 | 98.8% | 416.43 | C20H24N4O6 | 10 MG
SDP
Catalog No. 5000203324
Encompass_Preferred
MP-Ala-Ala-PAB is a peptide-based cleavable ADC linker used as an Fmoc-protected building block in the synthesis of antibody-drug conjugates (ADCs).
  • Cleavable valine-alanine-para-aminobenzyl (PAB) spacer for controlled payload release
  • Fmoc-protected N-terminus compatible with peptide and solid-phase synthesis
  • Supplied in small research pack sizes suitable for conjugation chemistry
  • High purity suitable for analytical and synthetic applications
Catalog No. 50-002-03324 Supplier Medchemexpress LLC Supplier No. HY14846310MG
MP-Ala-Ala-PAB is a peptide-based cleavable ADC linker used as an Fmoc-protected building block in the synthesis of antibody-drug conjugates (ADCs).
  • Cleavable valine-alanine-para-aminobenzyl (PAB) spacer for controlled payload release
  • Fmoc-protected N-terminus compatible with peptide and solid-phase synthesis
  • Supplied in small research pack sizes suitable for conjugation chemistry
  • High purity suitable for analytical and synthetic applications