Learn More
REFEYN INC Alignment Assist Package to affix the sample well cassette precisely and reliably to the sample carrier slide. This includes:
SDP
Supplier:  REFEYN INC MPCON21019
Encompass
Alignment Assist Package to affix the sample well cassette precisely and reliably to the sample carrier slide. This includes: - MP alignment tool - Carbon fibre tip tweezers- Set of magnets (4)
Catalog No. NC3737093