Learn More
REFEYN INC MP Auto - Alignment Kit, Package to affix the sample well cassette precisely and reliably to the sample carrier slide.
SDP
Catalog No. NC3737097
Encompass
Alignment Assist Package to affix the sample well cassette precisely and reliably to the sample carrier slide. This includes: - MP Auto alignment tool - Carbon fibre tip tweezers- Set of magnets (4)
Catalog No. NC3737097 Supplier REFEYN INC Supplier No. MPCON21020
Alignment Assist Package to affix the sample well cassette precisely and reliably to the sample carrier slide. This includes: - MP Auto alignment tool - Carbon fibre tip tweezers- Set of magnets (4)