Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 1X Phosphate Buffered Saline, Dulbecco's, without Magnesium and Calcium

Catalog No. MP1860454X6 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
6 x 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1860454X6 6 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP1860454X6 Supplier MP Biomedicals™ Supplier No. 091860454X6
Add to Cart
Add to Cart

Clear, colorless saline solution without magnesium and calcium

  • MDL Number: MFCD00131855
  • Format: Solution
  • pH: 7 to 7.6    
  • Osmolality: 270 to 305 mM/kg

For handling and culturing of mammalian cells

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Color Clear
Concentration 1 X
pH 7 to 7.6
Physical Form Liquid
Quantity 6 x 500 mL
Formulation Potassium Chloride 200mg/L, Potassium Phosphate Monobasic 200mg/L, Sodium Chloride 8000mg/L, Sodium Phosphate Dibasic 1150mg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.