Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ β2-Glycoprotein I

Catalog No. ICN19407580 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19407580 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19407580 Supplier MP Biomedicals™ Supplier No. 0219407580
Only null left
Add to Cart
Add to Cart

From human plasma

Apoliprotein H. Purified protein containing glycine and NaCl as stabilizers. Possible target of antiphospholipid antibodies and a component involved in phospholipid immunogenicity in the APA syndrome.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10mg/mL
Formulation The product is lyophilized from 0.01M Ammonium Bicarbonate
Molecular Weight (g/mol) 48
Name beta 2-Glycoprotein I from Human Plasma
Purification Method Proprietary Column Chromatography
Quantity 100 μg
Source Human Plasma
Storage Requirements -20°C
Form Lyophilized
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.