Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 7-Aminoactinomycin D

Catalog No. ICN19585601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19585601 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19585601 Supplier MP Biomedicals™ Supplier No. 0219585601

DNA intercalator and growth inhibitor against certain forms of leukemia and sarcomas

Purity: ≥95%. MW: 1270.4. Synonyms: 7-AAD, 7-Amino-AMD, 7-Actinomycin C1, 7-Aminodactinomycin.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Flow Cytometry
Quantity 1 mg
Purity ≥95%
Content And Storage 4°C
Product Type 7-Aminoactinomycin D
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.