Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Albumin 35%, Path 5

Catalog No. ICN810111 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN810111 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN810111 Supplier MP Biomedicals™ Supplier No. 08810111
Add to Cart
Add to Cart

Used as a medium in density gradient centrifugation or as an addition to tissue culture growth medium

Albumin bovine path 5 is a single polypeptide chain consisting of about 583 amino acid residues and no carbohydrates.

  • MDL Number: MFCD00130384
  • Format: Solution
  • Concentration: 35g/dl
  • Sterile albumins
  • Stable at least 2 years at 5°C

Used as an additive to cell culture media, especially serum-free media.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 34.8g/dL
For Use With (Application) Immunoassay, Mammalian Cell Culture
Name Bovine Albumin, Path 5
pH Range 7
Quantity 50 mL
Storage Requirements 2° to 8°C
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.