Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Albumin, Bovine, Fatty Acid-Free

Catalog No. ICN15240105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15240105 5 g
ICN15240150 50 g
ICN15240180 100 g
ICN15240110 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15240105 Supplier MP Biomedicals™ Supplier No. 0215240105
Add to Cart
Add to Cart

Prepared by Fraction V method

  • Low fatty acids
  • Purity: >98%
  • pH: 7

For best cell culture performance, see alternate Mfr. No. 180575

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Immunohematology, Serology
Molecular Weight (g/mol) 66.338
Name Bovine Albumin
pH Range 5.2 to 7
Quantity 5 g
Source Bovine
Storage Requirements 2° to 8°C
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.