Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Albumin, Bovine, For IVD, Low IgG

Catalog No. ICN810032 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50g
100g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN810032 50g
ICN810034 500g
ICN810033 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN810032 Supplier MP Biomedicals™ Supplier No. 08810032
Add to Cart
Add to Cart

Prepared by heat shock fractionation

  • For IVD applications, immunoassays, enzyme stabilization
  • Low protease, conductivity and visible absorbance
  • Purity: >98%
  • pH: 7

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Immunoassay, Mammalian Cell Culture
Molecular Weight (g/mol) ∽66
Name Bovine Blood Albumin Fraction V
pH Range 7
Quantity 50g
Source Bovine Blood
Storage Requirements 0° to 5°C
Form Crystalline
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.