Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Albumin Human 10%, Prepared from Fraction V human albumin

Catalog No. ICN810171 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN810171 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN810171 Supplier MP Biomedicals™ Supplier No. 08810171
Only null left
Add to Cart
Add to Cart

Supplied at a pH of 7.1 ±0.3 and contains 0.05% sodium azide as a preservative

  • CAS: 70024-90-7
  • Purity: >96.0 %

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 65
Name Human Albumin Protein
pH Range 7.1
Quantity 50 mL
Source Human Albumin
Storage Requirements 0° to 5°C
Form Liquid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.